Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Benzyloxypyridine-3-boronic acid pinacol ester, 95%

$350.96 - $350.96

Chemical Identifiers

Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid pinacol ester
CAS 832735-54-3
Molecular Formula C18H22BNO3
Formula Weight 311.19
MDL Number MFCD06798263
Synonym 2-Benzyloxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2770803

 
alfa aesar™
H2770803
1g Each for $350.96
Description & Specifications

Specifications

Formula Weight 311.19
Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid pinacol ester
Assay 95%
Molecular Formula C18H22BNO3
MDL Number MFCD06798263
CAS 832735-54-3
Synonym 2-Benzyloxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Melting Point 138° to 142°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.