missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Benzyloxypyridine-3-boronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445320010
Specifications
6-Benzyloxypyridine-3-boronic acid | |
107°C to 110°C | |
929250-35-1 | |
C12H12BNO3 | |
CUZOGAUUSKBDAO-UHFFFAOYSA-N | |
(6-phenylmethoxypyridin-3-yl)boronic acid | |
17750216 | |
97% |
Glass Bottle | |
1g | |
96% min. (HPLC) | |
2-benzyloxy pyridine-5-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxypyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxypyridine-3-boronic acid | |
B(C1=CN=C(C=C1)OCC2=CC=CC=C2)(O)O | |
229.04 | |
229.04 |
Chemical Identifiers
929250-35-1 | |
229.04 | |
2-benzyloxy pyridine-5-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxypyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxypyridine-3-boronic acid | |
(6-phenylmethoxypyridin-3-yl)boronic acid |
C12H12BNO3 | |
CUZOGAUUSKBDAO-UHFFFAOYSA-N | |
17750216 | |
B(C1=CN=C(C=C1)OCC2=CC=CC=C2)(O)O |
Safety and Handling
GHS Signal Word: Warning