Please login to your online account to display your discounted pricing

6-Benzyloxypyridine-3-boronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid
Assay Percent Range 97%
Molecular Formula C12H12BNO3
Formula Weight 229.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC445320010

 
thermo fisher scientific
AC445320010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445320050

 
thermo fisher scientific
AC445320050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.04
Quantity 1g
Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid
Assay Percent Range 97%
CAS 929250-35-1
Molecular Formula C12H12BNO3