6-Benzyloxypyridine-3-boronic acid, 97%, ACROS Organics™
Manufacturer:  Thermo Fisher Scientific AC445320050
 View more versions of this product
Catalog No. AC445320050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
6-Benzyloxypyridine-3-boronic acid
5g
96% min. (HPLC)
2-benzyloxy pyridine-5-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxypyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxypyridine-3-boronic acid
B(C1=CN=C(C=C1)OCC2=CC=CC=C2)(O)O
229.04
229.04
107°C to 110°C
929250-35-1
C12H12BNO3
CUZOGAUUSKBDAO-UHFFFAOYSA-N
(6-phenylmethoxypyridin-3-yl)boronic acid
17750216
97%
Chemical Identifiers
929250-35-1
229.04
2-benzyloxy pyridine-5-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxypyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxypyridine-3-boronic acid
(6-phenylmethoxypyridin-3-yl)boronic acid
C12H12BNO3
CUZOGAUUSKBDAO-UHFFFAOYSA-N
17750216
B(C1=CN=C(C=C1)OCC2=CC=CC=C2)(O)O
Safety and Handling
Safety and Handling
GHS Signal Word: Warning
SDS Documents