Please login to your online account to display your discounted pricing

6-Benzylaminopurine riboside, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Benzylaminopurine riboside
Name Note 99+%
CAS 4294-16-0
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C17H19N5O4
Formula Weight 357.36
MDL Number MFCD00005740
Synonym N6-Benzyladenosine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC226420010

  SDS
thermo fisher scientific
226420010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 357.36
Quantity 1g
Color White
EINECS Number 224-298-9
Beilstein 26, IV, 3682
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 6-Benzylaminopurine riboside
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C17H19N5O4
MDL Number MFCD00005740
CAS 4294-16-0
Synonym N6-Benzyladenosine
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic