Please login to your online account to display your discounted pricing

6-Benzylaminopurine, 99%, ACROS Organics™

$103.83 - $103.83

Chemical Identifiers

Chemical Name or Material 6-Benzylaminopurine
CAS 1214-39-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H11N5
Formula Weight 225.25
MDL Number MFCD00005572
Synonym BA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC226410250

  SDS
thermo fisher scientific
AC226410250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC226410050

  SDS
fisher scientific acros
226410050
Glass bottle 5g Each for $103.83

AC226410010

  SDS
fisher scientific acros
226410010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 225.25
Quantity 25g
Color White to very faint yellow
Melting Point 229° to 233°C
Beilstein 26, IV, 3575
CAS Max % 100.0
Loss on Drying 0.5% max.
Packaging Glass bottle
Chemical Name or Material 6-Benzylaminopurine
Assay Percent Range 99%
Molecular Formula C12H11N5
CAS 1214-39-7
CAS Min % 98.5
Health Hazard 1 Warning
MDL Number MFCD00005572
Infrared Spectrum Authentic
Synonym BA
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.