missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Benzylaminopurine, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 226410010

 View more versions of this product

Catalog No. AC226410010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
225.25
White to very faint yellow
229° to 233°C
Glass bottle
MFCD00005572
1214-39-7
BA
Authentic
Crystalline Powder
1g
26, IV, 3575
100.0
C12H11N5
99%
98.5
0.5% max.
6-Benzylaminopurine

Chemical Identifiers

1214-39-7
100.0
C12H11N5
MFCD00005572
6-Benzylaminopurine
98.5
99%
225.25
BA
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning