Learn More
6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals
$123.58 - $123.58
Chemical Identifiers
| CAS | 7472-43-7 |
|---|---|
| Molecular Formula | C13H16O3 |
| Molecular Weight (g/mol) | 220.27 |
| MDL Number | MFCD00039608 |
| InChI Key | DOQWHEUDAHLEPT-UHFFFAOYSA-N |
| Synonym | 6-benzoylhexanoic acid, 7-oxo-7-phenyl-heptanoic acid, 6-benzoyl hexanoic acid, benzeneheptanoic acid, z-oxo, 7-oxo-7-phenylheptanoicacid, #, benzeneheptanoic acid,, ae-oxo, alpha-oxo benzeneheptanoic acid, benzeneheptanoic acid,alpha-oxo |
| PubChem CID | 344830 |
| IUPAC Name | 7-oxo-7-phenylheptanoic acid |
| SMILES | OC(=O)CCCCCC(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0843103
|
Thermo Scientific Chemicals
L0843103 |
1 g |
Each for $123.58
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7472-43-7 | |
| 220.27 | |
| DOQWHEUDAHLEPT-UHFFFAOYSA-N | |
| 344830 | |
| OC(=O)CCCCCC(=O)C1=CC=CC=C1 |
| C13H16O3 | |
| MFCD00039608 | |
| 6-benzoylhexanoic acid, 7-oxo-7-phenyl-heptanoic acid, 6-benzoyl hexanoic acid, benzeneheptanoic acid, z-oxo, 7-oxo-7-phenylheptanoicacid, #, benzeneheptanoic acid,, ae-oxo, alpha-oxo benzeneheptanoic acid, benzeneheptanoic acid,alpha-oxo | |
| 7-oxo-7-phenylheptanoic acid |
Specifications
| 7472-43-7 | |
| C13H16O3 | |
| 1 g | |
| 6-benzoylhexanoic acid, 7-oxo-7-phenyl-heptanoic acid, 6-benzoyl hexanoic acid, benzeneheptanoic acid, z-oxo, 7-oxo-7-phenylheptanoicacid, #, benzeneheptanoic acid,, ae-oxo, alpha-oxo benzeneheptanoic acid, benzeneheptanoic acid,alpha-oxo | |
| OC(=O)CCCCCC(=O)C1=CC=CC=C1 | |
| 220.27 | |
| 220.27 | |
| 6-Benzoylhexanoic acid |
| 83°C to 86°C | |
| MFCD00039608 | |
| 2694641 | |
| DOQWHEUDAHLEPT-UHFFFAOYSA-N | |
| 7-oxo-7-phenylheptanoic acid | |
| 344830 | |
| 94% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only