missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Aminoquinoline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 369640250

 View more versions of this product

Catalog No. AC369640250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Aminoquinoline
115.0°C to 119.0°C
97.5% min. (HPLC)
C9H8N2
22, V,10, 303
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, benzene, diethyl ether,heptane
C1=CC2=C(C=CC(=C2)N)N=C1
144.18
144.18
580-15-4
192.0°C to 195.0°C (14.0 mmHg)
Glass bottle
25 g
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
RJSRSRITMWVIQT-UHFFFAOYSA-N
quinolin-6-amine
11373
98%

Chemical Identifiers

580-15-4
144.18
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
quinolin-6-amine
C9H8N2
RJSRSRITMWVIQT-UHFFFAOYSA-N
11373
C1=CC2=C(C=CC(=C2)N)N=C1
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 209-453-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only