missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminoquinoline, 98%

Manufacturer:  Alfa Aesar™ L1381914

 View more versions of this product

Catalog No. AAL1381914



Description

Description

Applications
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

Solubility
Soluble in methanol, chloroform, ethanol, and benzene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications

Specifications

6-Aminoquinoline
146°C (0.3mmHg)
25g
C9H8N2
113320
RJSRSRITMWVIQT-UHFFFAOYSA-N
quinolin-6-amine
11373
98%
Odorless
115°C to 119°C
580-15-4
MFCD00006803
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
C1=CC2=C(C=CC(=C2)N)N=C1
144.177
144.18

Chemical Identifiers

580-15-4
144.177
RJSRSRITMWVIQT-UHFFFAOYSA-N
11373
C1=CC2=C(C=CC(=C2)N)N=C1
C9H8N2
MFCD00006803
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
quinolin-6-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents