Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminopyridine-3-boronic acid pinacol ester, 97%

$167.78 - $538.74

Chemical Identifiers

Chemical Name or Material 6-Aminopyridine-3-boronic acid pinacol ester
CAS 827614-64-2
Molecular Formula C11H17BN2O2
Formula Weight 220.08
MDL Number MFCD05663837
Synonym 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2768503

 
alfa aesar™
H2768503
1g Each for $167.78

AAH2768506

 
alfa aesar™
H2768506
5g Each for $538.74
Description & Specifications

Specifications

Formula Weight 220.08
Chemical Name or Material 6-Aminopyridine-3-boronic acid pinacol ester
Melting Point 131° to 135°C
Quantity 1g
Assay 97%
Molecular Formula C11H17BN2O2
MDL Number MFCD05663837
CAS 827614-64-2
Synonym 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is an important raw material and intermediate. Substrate employed in a microwave-assisted, four-component coupling process leading to amino substituted imidazopyridines.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.