Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminopenicillanic acid, 98%

$58.31 - $870.91

Chemical Identifiers

Chemical Name or Material 6-Aminopenicillanic acid
CAS 551-16-6
Molecular Formula C8H12N2O3S
Formula Weight 216.26
MDL Number MFCD00005176
Beilstein 15080
Merck Index 14,458
Synonym 6-APA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1305609

  SDS
alfa aesar™
A1305609
10g Each for $58.31

AAA1305618

  SDS
alfa aesar™
A1305618
50g Each for $201.91

AAA1305630

  SDS
alfa aesar™
A1305630
250g Each for $870.91
Description & Specifications

Specifications

Formula Weight 216.26
Recommended Storage 2° to 8°C
MDL Number MFCD00005176
Optical Rotation +275° (c=1 in 0.1N HCl)
Synonym 6-APA
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Melting Point 200°C (dec.)
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 551-16-6
RTECS Number XH8225000
Chemical Name or Material 6-Aminopenicillanic acid
Quantity 10g
Assay 98%
Molecular Formula C8H12N2O3S
Merck Index 14,458
TSCA TSCA
Health Hazard 1 Danger
Beilstein 15080

Applications
6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.

Solubility
Soluble in water and hydrochoric acid.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

2° to 8°C