missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminonicotinic acid, 98%

Manufacturer:  Alfa Aesar™ A1122303

 View more versions of this product

Catalog No. AAA1122303



Specifications

Specifications

6-Aminonicotinic acid
~320°C (decomposition)
1g
MFCD00006326
14,456
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
6-aminopyridine-3-carboxylic acid
18496
138.13
3167-49-5
98%
C6H6N2O2
115992
6-Aminopyridine-3-carboxylic acid
C1=CC(=NC=C1C(=O)O)N
138.126
CHEBI:68583

Chemical Identifiers

3167-49-5
138.126
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
18496
6-aminopyridine-3-carboxylic acid
C6H6N2O2
MFCD00006326
6-Aminopyridine-3-carboxylic acid
CHEBI:68583
C1=CC(=NC=C1C(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents