Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminonicotinamidoxime, 97%

$298.82 - $298.82

Chemical Identifiers

Chemical Name or Material 6-Aminonicotinamidoxime
CAS 468068-28-2
Molecular Formula C6H8N4O
Formula Weight 152.16g/mol
MDL Number MFCD09910192
Synonym 6-Aminopyridine-3-carboxamidoxime; 5-(2-Aminopyridyl)amidoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5172003

 
alfa aesar™
H51720-03
Each for $298.82
Description & Specifications

Specifications

Melting Point 192° to 195°C
Quantity 1g
Chemical Name or Material 6-Aminonicotinamidoxime
CAS 468068-28-2
Assay 97%
Molecular Formula C6H8N4O
Formula Weight 152.16g/mol
MDL Number MFCD09910192
Synonym 6-Aminopyridine-3-carboxamidoxime; 5-(2-Aminopyridyl)amidoxime
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.