Learn More
6-Aminonicotinamide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0669206
Description
6-Aminonicotinamide induces apoptosis in tumor cells. It is clinically used in disseminated neoplastic disease. It also acts as 6-phosphogluconate dehydrogenase inhibitor. It aids in the treatment of psoriasis. It is used as cancer chemotherapeutic drug in animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in Dimethyl sulfoxide (DMSO). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store at -20°C
Specifications
6-Aminonicotinamide | |
246°C to 248°C | |
MFCD00006327 | |
116042 | |
Soluble in Dimethyl sulfoxide (DMSO). Insoluble in water. | |
NC(=O)C1=CC=C(N)N=C1 | |
137.14 | |
CHEBI:74514 | |
99% |
329-89-5 | |
C6H7N3O | |
5 g | |
6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid | |
ZLWYEPMDOUQDBW-UHFFFAOYSA-N | |
6-aminopyridine-3-carboxamide | |
9500 | |
137.14 |
Chemical Identifiers
329-89-5 | |
137.14 | |
ZLWYEPMDOUQDBW-UHFFFAOYSA-N | |
9500 | |
6-aminopyridine-3-carboxamide |
C6H7N3O | |
MFCD00006327 | |
6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid | |
CHEBI:74514 | |
NC(=O)C1=CC=C(N)N=C1 |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360
EINECSNumber : 206-349-7
RTECSNumber : US4550000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only