Please login to your online account to display your discounted pricing

6-Aminonicotinamide, 98%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC104150050

 View more versions of this product

Catalog No. AC104150050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 6-Aminonicotinamide
CAS 329-89-5
Packaging Glass bottle
Assay Percent Range 98%
Quantity 5g
Molecular Formula C6H7N3O
Formula Weight 137.4

Specifications

Chemical Name or Material 6-Aminonicotinamide
Assay Percent Range 98%
Molecular Formula C6H7N3O
Formula Weight 137.4