missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Aminonicotinamide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 104150010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
6-Aminonicotinamide | |
97.5% min. (HPLC) | |
C6H7N3O | |
MFCD00006327 | |
6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid | |
ZLWYEPMDOUQDBW-UHFFFAOYSA-N | |
6-aminopyridine-3-carboxamide | |
9500 | |
137.4 |
329-89-5 | |
Glass bottle | |
H2N(C5H3N)CONH2 | |
1g | |
Solubility in water: insoluble | |
NC(=O)C1=CC=C(N)N=C1 | |
137.14 | |
CHEBI:74514 | |
98% |
Chemical Identifiers
329-89-5 | |
137.14 | |
ZLWYEPMDOUQDBW-UHFFFAOYSA-N | |
9500 | |
6-aminopyridine-3-carboxamide |
C6H7N3O | |
MFCD00006327 | |
6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid | |
CHEBI:74514 | |
NC(=O)C1=CC=C(N)N=C1 |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
GHS P Statement
Use personal protective equipment as required.
IF exposed or concerned: Get medical attention/advice.
GHS Signal Word: Danger
EINECSNumber : 206-349-7
RUO â Research Use Only