Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminoindole, 98%

$258.61 - $258.61

Chemical Identifiers

Chemical Name or Material 6-Aminoindole
CAS 5318-27-4
Molecular Formula C8H8N2
Formula Weight 132.17g/mol
MDL Number MFCD02093959
Beilstein 112190
Synonym 6-Indolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL17793ME

 
alfa aesar™
L17793-ME
500mg Each for $258.61
Description & Specifications

Specifications

Melting Point 70° to 76°C
Boiling Point 161° to 166°C (2mmHg)
Quantity 500mg
Chemical Name or Material 6-Aminoindole
CAS 5318-27-4
Assay 98%
Molecular Formula C8H8N2
Formula Weight 132.17g/mol
MDL Number MFCD02093959
Beilstein 112190
Synonym 6-Indolamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C