missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Aminoindole, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC433330010

 View more versions of this product

Catalog No. AC433330010



Specifications

Specifications

6-Aminoindole
Glass bottle
White to Brown
1g
96.0
97%
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
1H-indol-6-amine
256096
Crystalline Powder
Authentic
161.0°C to 166.0°C (2.3 Torr)
66.0°C to 68.0°C
5318-27-4
100.0
C8H8N2
6-aminoindole, 6-amino-1h-indole, 6-amino indole, 1h-indol-6-ylamine, indole-6-ylamine, 1h-indol-6-amin, 6-indolamine, indol-6-amine, pubchem7311, 6-aminoindole
C1=CC(=CC2=C1C=CN2)N
132.166
132.17
97%

Chemical Identifiers

5318-27-4
132.166
MIMYTSWNVBMNRH-UHFFFAOYSA-N
256096
C1=CC(=CC2=C1C=CN2)N
C8H8N2
MFCD02093959
6-aminoindole, 6-amino-1h-indole, 6-amino indole, 1h-indol-6-ylamine, indole-6-ylamine, 1h-indol-6-amin, 6-indolamine, indol-6-amine, pubchem7311, 6-aminoindole
1H-indol-6-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents