Please login to your online account to display your discounted pricing

6-Aminoindole, 97%, ACROS Organics™

$122.70 - $122.70

Chemical Identifiers

Chemical Name or Material 6-Aminoindole
CAS 5318-27-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8N2
Formula Weight 132.17
MDL Number MFCD02093959
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC433330050

  SDS
thermo fisher scientific
AC433330050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC433330010

  SDS
thermo fisher scientific
AC433330010
Glass bottle 1g Each for $122.70
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 132.17
Physical Form Crystalline Powder
Quantity 5g
Color White to brown
Melting Point 66° to 70°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 5318-27-4
Molecular Formula C8H8N2
Chemical Name or Material 6-Aminoindole
Assay Percent Range 97%
MDL Number MFCD02093959
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.