missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Aminoindan-1-one, 97%, ACROS Organics™

Chemical Identifiers

CAS 69975-65-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00100747
Chemical Name or Material 6-Aminoindan-1-one
Assay ≥96.0% (HPLC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC432830050 SDS Thermo Fisher Scientific
AC432830050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432830010 SDS Thermo Fisher Scientific
AC432830010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

69975-65-1
100.0
C9H9NO
MFCD00100747
≥96.0% (HPLC)
96.0
97%
147.18
6-Aminoindan-1-one

Specifications

Solution
5g
155° to 159°C
96.0
C9H9NO
MFCD00100747
Glass bottle
≥96.0% (HPLC)
147.18
Light brown to black
100.0
Authentic
97%
69975-65-1
6-Aminoindan-1-one

Safety and Handling

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning