Please login to your online account to display your discounted pricing

6-Aminoindan-1-one, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC432830050

 View more versions of this product

Catalog No. AC432830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Assay ≥96.0% (HPLC)
Formula Weight 147.18
Quantity 5g
Color Light brown to black
Melting Point 155° to 159°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C9H9NO
Infrared Spectrum Authentic
Assay Percent Range 97%
Chemical Name or Material 6-Aminoindan-1-one
MDL Number MFCD00100747
CAS 69975-65-1
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 6-Aminoindan-1-one
CAS 69975-65-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00100747
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents