Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Aminobenzoxazole, 95%

Chemical Identifiers

Chemical Name or Material 6-Aminobenzoxazole
CAS 177492-52-3
Formula Weight 134.1
MDL Number MFCD03791187
Synonym Benzoxazol-6-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3448503

 
alfa aesar™
H3448503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3448506

 
alfa aesar™
H3448506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH34485MD

 
alfa aesar™
H34485MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 95° to 97°C
Formula Weight 134.1
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 6-Aminobenzoxazole
Physical Form Solid
Assay 95%
Color Orange to brown
Quantity 1g
MDL Number MFCD03791187
CAS 177492-52-3
Sensitivity Air Sensitive
Synonym Benzoxazol-6-amine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1 2° to 8°C

Applications
6-Aminobenzoxazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C