Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-7-deazapurine hydrogen sulfate, 97%

Chemical Identifiers

Chemical Name or Material 6-Amino-7-deazapurine hydrogen sulfate
CAS 769951-32-8
Formula Weight 232.21
MDL Number MFCD09787504
Synonym 4-Amino-7H-pyrrolo[2,3-d]pyrimidine hydrogen sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6460203

  SDS
alfa aesar™
H6460203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6460206

  SDS
alfa aesar™
H6460206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64602MD

  SDS
alfa aesar™
H64602MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.21
Physical Form Powder
Color Brown
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4-Amino-7H-pyrrolo[2,3-d]pyrimidine hydrogen sulfate
Assay 97%
CAS 769951-32-8
Storage Note 1 -30° to -10°C
Chemical Name or Material 6-Amino-7-deazapurine hydrogen sulfate
MDL Number MFCD09787504
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C