Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-7-deazapurine, 97%

Chemical Identifiers

Chemical Name or Material 6-Amino-7-deazapurine
CAS 1500-85-2
Formula Weight 134.14
MDL Number MFCD01686848
Synonym 7-Deazaadenine; 7H-Pyrrolo[2,3-d]pyrimidin-4-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6698703

  SDS
alfa aesar™
H6698703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6698706

  SDS
alfa aesar™
H6698706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 257° to 262°C
Formula Weight 134.14
Chemical Name or Material 6-Amino-7-deazapurine
CAS 1500-85-2
Health Hazard 1 Warning
Assay 97%
Quantity 1g
Sensitivity Moisture & Light Sensitive
MDL Number MFCD01686848
Synonym 7-Deazaadenine; 7H-Pyrrolo[2,3-d]pyrimidin-4-amine
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to pale brown
Physical Form Solid
Storage Note 1 2° to 8°C
RTECS Number UY8850000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C