missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Amino-3-nitro-2-picoline, 98%, ACROS Organics™

$141.55 - $558.30

Chemical Identifiers

CAS 22280-62-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H7N3O2
Formula Weight 153
MDL Number MFCD00053582
Synonym 2-Amino-6-methyl-5-nitropyridine
Chemical Name or Material 6-Amino-3-nitro-2-picoline
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC360770050 SDS Thermo Fisher Scientific
AC360770050
Glass bottle 5g Each for $558.30
AC360770010 SDS Thermo Fisher Scientific
AC360770010
Glass bottle 1g Each for $141.55
Specifications Description & Specifications

Chemical Identifiers

22280-62-2
100.0
C6H7N3O2
MFCD00053582
6-Amino-3-nitro-2-picoline
97.5
98%
153
2-Amino-6-methyl-5-nitropyridine
≥97.5%

Specifications

153
Powder
186° to 192°C
C6H7N3O2
22280-62-2
97.5
2-Amino-6-methyl-5-nitropyridine
6-Amino-3-nitro-2-picoline
Beige to yellow-brown
5g
100.0
Authentic
98%
MFCD00053582
Glass bottle
≥97.5%

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning