Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-3-iodo-2-methylpyridine, 95%

$113.01 - $253.14

Chemical Identifiers

Chemical Name or Material 6-Amino-3-iodo-2-methylpyridine
CAS 75073-11-9
Molecular Formula C6H7IN2
Formula Weight 234.04
MDL Number MFCD04138418
Synonym 6-Amino-3-iodo-2-picoline; 5-Iodo-6-methyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2726503

  SDS
alfa aesar™
H2726503
1g Each for $253.14

AAH27265MD

  SDS
alfa aesar™
H27265MD
250mg Each for $113.01
Description & Specifications

Specifications

Formula Weight 234.04
Chemical Name or Material 6-Amino-3-iodo-2-methylpyridine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
CAS 75073-11-9
Synonym 6-Amino-3-iodo-2-picoline; 5-Iodo-6-methyl-2-pyridinamine
Assay 95%
Quantity 1g
Molecular Formula C6H7IN2
MDL Number MFCD04138418
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.