missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Amino-2-pyridinemethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 450040010
Specifications
6-Amino-2-pyridinemethanol | |
Authentic | |
Glass Bottle | |
1g | |
YGMPMXTWKROXPP-UHFFFAOYSA-N | |
(6-aminopyridin-2-yl)methanol | |
11182606 | |
97% |
79651-64-2 | |
96% min. (GC) | |
C6H8N2O | |
6-aminopyridin-2-yl methanol, 6-amino-2-pyridinemethanol, 2-aminopyridine-6-methanol, 6-amino-pyridin-2-yl-methanol, 2-pyridinemethanol, 6-amino, 2-amino-6-hydroxymethylpyridine, 6-aminopyridine-2-methanol, 6-amino-2-pyridinyl methanol, 2-amino-6-hydroxymethyl pyridine, 2-hydroxymethyl-6-aminopyridine | |
C1=CC(=NC(=C1)N)CO | |
124.14 | |
124.14 |
Chemical Identifiers
79651-64-2 | |
124.14 | |
6-aminopyridin-2-yl methanol, 6-amino-2-pyridinemethanol, 2-aminopyridine-6-methanol, 6-amino-pyridin-2-yl-methanol, 2-pyridinemethanol, 6-amino, 2-amino-6-hydroxymethylpyridine, 6-aminopyridine-2-methanol, 6-amino-2-pyridinyl methanol, 2-amino-6-hydroxymethyl pyridine, 2-hydroxymethyl-6-aminopyridine | |
(6-aminopyridin-2-yl)methanol |
C6H8N2O | |
YGMPMXTWKROXPP-UHFFFAOYSA-N | |
11182606 | |
C1=CC(=NC(=C1)N)CO |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only