missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Amino-2-naphthoic acid, 90%, ACROS Organics™

$245.51 - $245.51

Chemical Identifiers

CAS 116668-47-4
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.2
MDL Number MFCD01861831
InChI Key NZTPZUIIYNYZKT-UHFFFAOYSA-N
Synonym 2-amino-6-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-carboxynaphthalene, 6-amino-2-naphthoic, 6-amino-2-naphthoic acid, 6-amino-2-naphthoicacid, 6-amino-2-naphtoic acid, 6-aminoisonaphthoic acid, bidd:gt0289
PubChem CID 2733954
IUPAC Name 6-aminonaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC344500050
SDS View Documents
Thermo Fisher Scientific
344500050
5g Glass bottle
Each for $245.51
 
Description

Description

Specifications

Chemical Identifiers

116668-47-4
187.2
NZTPZUIIYNYZKT-UHFFFAOYSA-N
2733954
C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O
C11H9NO2
MFCD01861831
2-amino-6-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-carboxynaphthalene, 6-amino-2-naphthoic, 6-amino-2-naphthoic acid, 6-amino-2-naphthoicacid, 6-amino-2-naphtoic acid, 6-aminoisonaphthoic acid, bidd:gt0289
6-aminonaphthalene-2-carboxylic acid

Specifications

Authentic
Beige-Green
5g
88.0
88% min. (HPLC)
MFCD01861831
NZTPZUIIYNYZKT-UHFFFAOYSA-N
6-aminonaphthalene-2-carboxylic acid
2733954
Powder
6-Amino-2-naphthoic acid, 90%
Glass bottle
222°C to 227°C
116668-47-4
100.0
C11H9NO2
2-amino-6-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-carboxynaphthalene, 6-amino-2-naphthoic, 6-amino-2-naphthoic acid, 6-amino-2-naphthoicacid, 6-amino-2-naphtoic acid, 6-aminoisonaphthoic acid, bidd:gt0289
C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O
187.2
187.2
90%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS