Learn More
6-Amino-2-methylquinoline, 97%, Thermo Scientific Chemicals
$90.75 - $1060.21
Chemical Identifiers
CAS | 65079-19-8 |
---|---|
Molecular Formula | C10H10N2 |
Molecular Weight (g/mol) | 158.204 |
MDL Number | MFCD00052600 |
InChI Key | TYJFYUVDUUACKX-UHFFFAOYSA-N |
PubChem CID | 103148 |
IUPAC Name | 2-methylquinolin-6-amine |
SMILES | CC1=NC2=C(C=C1)C=C(C=C2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA13967MD
|
Thermo Scientific Chemicals
A13967MD |
250 mg |
Each for $90.75
|
|
|||||
AAA1396703
|
Thermo Scientific Chemicals
A1396703 |
1 g |
Each for $242.08
|
|
|||||
AAA1396706
|
Thermo Scientific Chemicals
A1396706 |
5 g |
Each for $1,060.21
|
|
|||||
Description
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
65079-19-8 | |
158.204 | |
TYJFYUVDUUACKX-UHFFFAOYSA-N | |
2-methylquinolin-6-amine |
C10H10N2 | |
MFCD00052600 | |
103148 | |
CC1=NC2=C(C=C1)C=C(C=C2)N |
Specifications
65079-19-8 | |
C10H10N2 | |
250 mg | |
6-amino-2-methylquinoline, 2-methyl-6-quinolinamine, 6-quinolinamine, 2-methyl, 6-aminoquinaldine, 2-methyl-6-aminoquinoline, 6-quinolinamine,2-methyl, 2-methyl-quinolin-6-ylamine, 2-methyl-6-quinolylamine, mqn, 2-methylquinoline-6-amine | |
TYJFYUVDUUACKX-UHFFFAOYSA-N | |
2-methylquinolin-6-amine | |
103148 | |
97% |
189°C to 191°C | |
MFCD00052600 | |
114795 | |
Soluble in water. | |
CC1=NC2=C(C=C1)C=C(C=C2)N | |
158.204 | |
158.2 | |
6-Amino-2-methylquinoline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 000-000-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only