missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Amino-2-methylnicotinonitrile, 97%, Maybridge

Manufacturer:  Fisher Scientific MO01041DA

 View more versions of this product

Catalog No. MO01041DA



Specifications

Specifications

6-Amino-2-methylnicotinonitrile
1g
97.0
C7H7N3
6-amino-2-methylnicotinonitrile, 6-amino-3-cyano-2-methylpyridine, 2-amino-5-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-amino-2-methyl, 6-amino-2-methyl-nicotinonitrile, pubchem14519, acmc-20ahz0, 6-amino-2-methyl-pyridine-3-carbonitrile, 6-amino-2-methyl nicotinonitrile, 2-methyl-3-cyano-6-amino-pyridine
CC1=C(C=CC(=N1)N)C#N
133.154
133.15
Amber Glass Bottle
183428-90-2
97%
MFCD05865116
OZAIJUZOQOBQDW-UHFFFAOYSA-N
6-amino-2-methylpyridine-3-carbonitrile
2794662
97%

Chemical Identifiers

183428-90-2
133.154
OZAIJUZOQOBQDW-UHFFFAOYSA-N
2794662
CC1=C(C=CC(=N1)N)C#N
C7H7N3
MFCD05865116
6-amino-2-methylnicotinonitrile, 6-amino-3-cyano-2-methylpyridine, 2-amino-5-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-amino-2-methyl, 6-amino-2-methyl-nicotinonitrile, pubchem14519, acmc-20ahz0, 6-amino-2-methyl-pyridine-3-carbonitrile, 6-amino-2-methyl nicotinonitrile, 2-methyl-3-cyano-6-amino-pyridine
6-amino-2-methylpyridine-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents