missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Amino-2-methylnicotinonitrile, 97%, Maybridge

$92.17 - $92.17

Chemical Identifiers

CAS 183428-90-2
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C7H7N3
Formula Weight 133.15
MDL Number MFCD05865116
Synonym 6-Amino-2-methylnicotinonitrile (IUPAC)
Chemical Name or Material 6-Amino-2-methylnicotinonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO01041DA SDS Fisher Scientific Uk Ltd
MO01041DA
Amber Glass Bottle 1g Each for $92.17
Specifications Description & Specifications

Chemical Identifiers

183428-90-2
97%
133.15
6-Amino-2-methylnicotinonitrile (IUPAC)
97%
97.0
C7H7N3
MFCD05865116
6-Amino-2-methylnicotinonitrile

Specifications

183428-90-2
Amber Glass Bottle
97%
133.15
6-Amino-2-methylnicotinonitrile (IUPAC)
97%
97.0
1g
C7H7N3
MFCD05865116
6-Amino-2-methylnicotinonitrile

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning