Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-2-methylbenzothiazole, 99%

$62.49 - $207.80

Chemical Identifiers

Chemical Name or Material 6-Amino-2-methylbenzothiazole
CAS 2941-62-0
Molecular Formula C8H8N2S
Formula Weight 164.22
MDL Number MFCD00160076
Synonym 2-Methylbenzothiazol-6-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5031903

  SDS
alfa aesar™
H5031903
1g Each for $207.80

AAH50319MD

  SDS
alfa aesar™
H50319MD
250mg Each for $62.49
Description & Specifications

Specifications

Formula Weight 164.22
Chemical Name or Material 6-Amino-2-methylbenzothiazole
Assay 99%
Molecular Formula C8H8N2S
MDL Number MFCD00160076
CAS 2941-62-0
Synonym 2-Methylbenzothiazol-6-amine
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.