Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-2-methyl-3-nitropyridine, 95%

Chemical Identifiers

Chemical Name or Material 6-Amino-2-methyl-3-nitropyridine
CAS 22280-62-2
Formula Weight 153.14
MDL Number MFCD00053582
Synonym 2-Amino-6-methyl-5-nitropyridine; 6-Amino-3-nitro-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6121803

  SDS
alfa aesar™
H6121803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6121806

  SDS
alfa aesar™
H6121806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.14
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 6-Amino-2-methyl-3-nitropyridine
Color Yellow
Physical Form Powder
Assay 95%
Quantity 1g
MDL Number MFCD00053582
CAS 22280-62-2
Synonym 2-Amino-6-methyl-5-nitropyridine; 6-Amino-3-nitro-2-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1 -30° to -10°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C