Please login to your online account to display your discounted pricing

6-Amino-1H-indazole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Amino-1H-indazole
CAS 6967-12-0
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C7H7N3
Formula Weight 133.15g/mol
MDL Number MFCD00005696
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC430130250

  SDS
thermo fisher scientific
430130250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 204° to 210°C
Color Tan
Quantity 25g
Chemical Name or Material 6-Amino-1H-indazole
CAS 6967-12-0
CAS Min % 96.0
CAS Max % 100.0
Physical Form Tan powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C7H7N3
Formula Weight 133.15g/mol
MDL Number MFCD00005696
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.