Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-1-methyluracil, 97%

$54.78 - $180.09

Chemical Identifiers

Chemical Name or Material 6-Amino-1-methyluracil
CAS 2434-53-9
Molecular Formula C5H7N3O2
Formula Weight 141.13
MDL Number MFCD00075366
Beilstein 127771
Synonym 6-Amino-1-methyl-2,4(1H,3H)-pyrimidinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2544814

 
alfa aesar™
B2544814
25g Each for $54.78

AAB2544822

 
alfa aesar™
B2544822
100g Each for $180.09
Description & Specifications

Specifications

Formula Weight 141.13
Chemical Name or Material 6-Amino-1-methyluracil
Assay 97%
Molecular Formula C5H7N3O2
MDL Number MFCD00075366
CAS 2434-53-9
Beilstein 127771
Synonym 6-Amino-1-methyl-2,4(1H,3H)-pyrimidinedione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point >300°C
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless

Applications
6-Amino-1-methyluracil may be used in the preparation of 1,1′-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5′-pyrrolo[2,3-d]pyrimidine]-2,2′,4,4′,6′(1′H,3H,3′H,7′H,1′H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.

Solubility
Soluble in diluted sodium hydroxide solution.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.