missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Amino-1-hexanol, 94%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 294600100

 View more versions of this product

Catalog No. AC294600100



Description

Description

Specifications

Specifications

6-Amino-1-hexanol
Authentic
0.5% max.
Yellow to Brown
10g
92.0
92% min. (GC)
H2N(CH2)6OH
04, II, 748
Solubility in water: soluble
C(CCCO)CCN
117.19
117.19
94%
133°C
Glass bottle
233.0°C
53°C to 59°C
4048-33-3
100.0
C6H15NO
MFCD00008241
1-amino-6-hexanol, 1-hexanol, 6-amino, 6-amino-1-hexanol, 6-amino-hexan-1-ol, 6-aminohexanol, 6-hydroxyhexylamine, amidohexylalkohol, amidohexylalkohol german, aminohexyl alcohol, hexanol, 6-amino
SUTWPJHCRAITLU-UHFFFAOYSA-N
6-aminohexan-1-ol
19960
Crystals

Chemical Identifiers

4048-33-3
117.19
SUTWPJHCRAITLU-UHFFFAOYSA-N
19960
C(CCCO)CCN
C6H15NO
MFCD00008241
1-amino-6-hexanol, 1-hexanol, 6-amino, 6-amino-1-hexanol, 6-amino-hexan-1-ol, 6-aminohexanol, 6-hydroxyhexylamine, amidohexylalkohol, amidohexylalkohol german, aminohexyl alcohol, hexanol, 6-amino
6-aminohexan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 223-748-1

SDS Documents