Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Amino-1,2,3,4-tetrahydroisoquinoline, 97%

Chemical Identifiers

Chemical Name or Material 6-Amino-1,2,3,4-tetrahydroisoquinoline
CAS 72299-67-3
Formula Weight 148.21
MDL Number MFCD04114859
Beilstein 8480377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66983MD

  SDS
alfa aesar™
H66983MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.21
MDL Number MFCD04114859
CAS 72299-67-3
Color Off-white
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 6-Amino-1,2,3,4-tetrahydroisoquinoline
Quantity 250mg
Assay 97%
Beilstein 8480377
Storage Note 1 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C