Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one, 98%

$69.46 - $207.93

Chemical Identifiers

Chemical Name or Material 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one
CAS 26518-71-8
Molecular Formula C10H9NO3
Formula Weight 191.19
MDL Number MFCD01321313
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2740706

  SDS
alfa aesar™
H2740706
5g Each for $69.46

AAH2740714

  SDS
alfa aesar™
H2740714
25g Each for $207.93
Description & Specifications

Specifications

Formula Weight 191.19
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD01321313
Molecular Formula C10H9NO3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 26518-71-8
Health Hazard 1 Warning
Melting Point 193° to 199°C
Chemical Name or Material 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.