Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Acetyl-1,4-benzodioxane, 98%

$65.09 - $822.69

Chemical Identifiers

Chemical Name or Material 6-Acetyl-1,4-benzodioxane
CAS 2879-20-1
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00006823
Beilstein 164125
Synonym 1,4-Benzodioxan-6-yl methyl ketone; 3',4'-Ethylenedioxyacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1550606

  SDS
alfa aesar™
A1550606
5g Each for $65.09

AAA1550614

  SDS
alfa aesar™
A1550614
25g Each for $233.55

AAA1550622

  SDS
alfa aesar™
A1550622
100g Each for $822.69
Description & Specifications

Specifications

Formula Weight 178.19
Chemical Name or Material 6-Acetyl-1,4-benzodioxane
Assay 98%
Molecular Formula C10H10O3
MDL Number MFCD00006823
CAS 2879-20-1
Beilstein 164125
Synonym 1,4-Benzodioxan-6-yl methyl ketone; 3',4'-Ethylenedioxyacetophenone
Melting Point 81° to 84°C
Quantity 5g