Alfa Aesar™ 6-Acetyl-1,4-benzodioxane, 98%
Manufacturer:  Alfa Aesar™ A1550606
 View more versions of this product
Catalog No. AAA1550606
Specifications
Specifications
6-Acetyl-1,4-benzodioxane
5g
C10H10O3
164125
HGVWMTAIIYNQSI-UHFFFAOYSA-N
1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
76143
98%
81°C to 84°C
2879-20-1
MFCD00006823
6-acetyl-1,4-benzodioxane, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 1,4-benzodioxan-6-yl methyl ketone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 6-acetyl-1,4-benzodioxan, ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl, ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one, 6-acetyl-2h,3h-benzo e 1,4-dioxin
CC(=O)C1=CC2=C(C=C1)OCCO2
178.187
178.19
Chemical Identifiers
2879-20-1
178.187
HGVWMTAIIYNQSI-UHFFFAOYSA-N
76143
CC(=O)C1=CC2=C(C=C1)OCCO2
C10H10O3
MFCD00006823
6-acetyl-1,4-benzodioxane, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 1,4-benzodioxan-6-yl methyl ketone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 6-acetyl-1,4-benzodioxan, ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl, ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one, 6-acetyl-2h,3h-benzo e 1,4-dioxin
1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
Documents