Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Acetoxy-2-naphthoic acid, 98+%


Quantity 1g

Manufacturer: alfa aesar™  L1562003

 View more versions of this product

Catalog No. AAL1562003

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 230.22
Chemical Name or Material 6-Acetoxy-2-naphthoic acid
CAS 17295-26-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay ≥98%
Molecular Formula C13H10O4
MDL Number MFCD00059556
Beilstein 3310348
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 218° to 224°C
Quantity 1g

Specifications

Chemical Name or Material 6-Acetoxy-2-naphthoic acid
CAS 17295-26-0
Molecular Formula C13H10O4
Formula Weight 230.22
MDL Number MFCD00059556
Beilstein 3310348
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.