missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Acetoxy-2-naphthoic acid, 98+%

Manufacturer:  Alfa Aesar™ L1562006

 View more versions of this product

Catalog No. AAL1562006



Specifications

Specifications

6-Acetoxy-2-naphthoic acid
5g
C13H10O4
3310348
NFTLBCXRDNIJMI-UHFFFAOYSA-N
6-acetyloxynaphthalene-2-carboxylic acid
87038
≥98%
218°C to 224°C
17295-26-0
MFCD00059556
6-acetoxy-2-naphthoic acid, 6-acetyloxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-acetyloxy, 6-acetyloxy naphthalene-2-carboxylic acid, 6-hydroxy-2-naphthoic acid acetate, 6-acetoxy-2-naphthoicacid, acmc-209e5j, 6-acetoxy-2-naphtoic acid, 2-acetoxy-6-naphthoic acid, 6-acetyloxy-2-naphthoic acid
CC(=O)OC1=CC2=C(C=C1)C=C(C=C2)C(=O)O
230.219
230.22

Chemical Identifiers

17295-26-0
230.219
NFTLBCXRDNIJMI-UHFFFAOYSA-N
87038
CC(=O)OC1=CC2=C(C=C1)C=C(C=C2)C(=O)O
C13H10O4
MFCD00059556
6-acetoxy-2-naphthoic acid, 6-acetyloxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-acetyloxy, 6-acetyloxy naphthalene-2-carboxylic acid, 6-hydroxy-2-naphthoic acid acetate, 6-acetoxy-2-naphthoicacid, acmc-209e5j, 6-acetoxy-2-naphtoic acid, 2-acetoxy-6-naphthoic acid, 6-acetyloxy-2-naphthoic acid
6-acetyloxynaphthalene-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents