Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Acetamidohexanoic acid hydrate, 99%

$34.49 - $79.58

Chemical Identifiers

Chemical Name or Material 6-Acetamidohexanoic acid hydrate
CAS 57-08-9
Molecular Formula C8H15NO3·xH2O
Linear Formula CH3CONH(CH2)5CO2H·xH2O
Formula Weight 173.21 (Anhydrous)
MDL Number MFCD00004424
Merck Index 14,45
Synonym 6-Acetamidocaproic acid hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2362306

 
alfa aesar™
B2362306
5g Each for $34.49

AAB2362314

 
alfa aesar™
B2362314
25g Each for $79.58
Description & Specifications

Specifications

Formula Weight 173.21 (Anhydrous)
Chemical Name or Material 6-Acetamidohexanoic acid hydrate
Assay 99%
Molecular Formula C8H15NO3·xH2O
Linear Formula CH3CONH(CH2)5CO2H·xH2O
MDL Number MFCD00004424
CAS 57-08-9
Merck Index 14,45
Synonym 6-Acetamidocaproic acid hydrate
Melting Point 102° to 106°C
Quantity 5g