Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,8-Dihydroxy-2-(methylthio)purine, 98%

$42.34 - $162.73

Chemical Identifiers

Chemical Name or Material 6,8-Dihydroxy-2-(methylthio)purine
CAS 14443-37-9
Molecular Formula C6H6N4O2S
Formula Weight 198.21
MDL Number MFCD00022637
Synonym 6,8-Dihydroxy-2-(methylmercapto)purine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1726203

  SDS
alfa aesar™
A1726203
1g Each for $42.34

AAA1726206

  SDS
alfa aesar™
A1726206
5g Each for $162.73
Description & Specifications

Specifications

Formula Weight 198.21
Synonym 6,8-Dihydroxy-2-(methylmercapto)purine
MDL Number MFCD00022637
Melting Point >300°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C6H6N4O2S
Assay 98%
Chemical Name or Material 6,8-Dihydroxy-2-(methylthio)purine
CAS 14443-37-9
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.