Please login to your online account to display your discounted pricing

6,8-Dichloro-3-formylchromone, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6, 8-Dichloro-3-formylchromone
CAS 64481-10-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H4Cl2O3
Formula Weight 243.05
MDL Number MFCD00051510
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372830050

  SDS
thermo fisher scientific
AC372830050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 243.05
Quantity 5g
Color Light yellow
Melting Point 169° to 174°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 6, 8-Dichloro-3-formylchromone
Assay Percent Range 97%
Molecular Formula C10H4Cl2O3
MDL Number MFCD00051510
CAS 64481-10-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.