missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6,7-Dinitro-1,4-dihydroquinoxaline-2,3-dione

Potent competitive non-NMDA glutamate receptor antagonist

$57.20 - $169.64

Chemical Identifiers

CAS 2379-57-9
Molecular Formula C8H4N4O6
Molecular Weight (g/mol) 252.142
MDL Number MFCD00069257
InChI Key RWVIMCIPOAXUDG-UHFFFAOYSA-N
Synonym dnqx, unii-62t278s1mx, 6,7-dinitroquinoxaline-2,3-dione dnqx, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dinitro, 6,7-dinitroquinoxaline-2,3-diol, 6,7-dinitro-1,4-dihydro-quinoxaline-2,3-dione, 6,7-dinitroquinoxaline-2,3 1h,4h-dione, 1ftl, tocris-0189, lopac-d-0540
PubChem CID 3899541
IUPAC Name 6,7-dinitro-1,4-dihydroquinoxaline-2,3-dione
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])[N+](=O)[O-])NC(=O)C(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60911MA View Documents Alfa Aesar™
J60911MA
10mg Each for $57.20
 
AAJ60911MF View Documents Alfa Aesar™
J60911MF
50mg Each for $169.64
 
Specifications

Chemical Identifiers

2379-57-9
252.142
RWVIMCIPOAXUDG-UHFFFAOYSA-N
3899541
C1=C2C(=CC(=C1[N+](=O)[O-])[N+](=O)[O-])NC(=O)C(=O)N2
C8H4N4O6
MFCD00069257
dnqx, unii-62t278s1mx, 6,7-dinitroquinoxaline-2,3-dione dnqx, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dinitro, 6,7-dinitroquinoxaline-2,3-diol, 6,7-dinitro-1,4-dihydro-quinoxaline-2,3-dione, 6,7-dinitroquinoxaline-2,3 1h,4h-dione, 1ftl, tocris-0189, lopac-d-0540
6,7-dinitro-1,4-dihydroquinoxaline-2,3-dione

Specifications

Yellow
10mg
C8H4N4O6
dnqx, unii-62t278s1mx, 6,7-dinitroquinoxaline-2,3-dione dnqx, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dinitro, 6,7-dinitroquinoxaline-2,3-diol, 6,7-dinitro-1,4-dihydro-quinoxaline-2,3-dione, 6,7-dinitroquinoxaline-2,3 1h,4h-dione, 1ftl, tocris-0189, lopac-d-0540
RWVIMCIPOAXUDG-UHFFFAOYSA-N
6,7-dinitro-1,4-dihydroquinoxaline-2,3-dione
3899541
Powder
>300°C
2379-57-9
MFCD00069257
Soluble in DMSO
C1=C2C(=CC(=C1[N+](=O)[O-])[N+](=O)[O-])NC(=O)C(=O)N2
252.142
252.14
6,7-Dinitro-1,4-dihydroquinoxaline-2,3-dione
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C