missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals

$106.30 - $350.95

Chemical Identifiers

CAS 20232-39-7
Molecular Formula C11H14ClNO2
Molecular Weight (g/mol) 227.69
MDL Number MFCD11099370
InChI Key PQXVEYYRJHMTEV-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
PubChem CID 2724664
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline;hydrochloride
SMILES Cl.COC1=CC2=C(C=NCC2)C=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6413006
View Documents Promotion Details
Thermo Scientific Chemicals
H6413006
5 g
Each for $106.30
 
AAH6413014
View Documents Promotion Details
Thermo Scientific Chemicals
H6413014
25 g
Each for $350.95
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20232-39-7
227.69
PQXVEYYRJHMTEV-UHFFFAOYSA-N
2724664
Cl.COC1=CC2=C(C=NCC2)C=C1OC
C11H14ClNO2
MFCD11099370
6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
6,7-dimethoxy-3,4-dihydroisoquinoline;hydrochloride

Specifications

20232-39-7
Yellow
C11H14ClNO2
5 g
6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
Cl.COC1=CC2=C(C=NCC2)C=C1OC
227.69
227.69
Solid
198°C to 202°C
Odorless
MFCD11099370
3712033
PQXVEYYRJHMTEV-UHFFFAOYSA-N
6,7-dimethoxy-3,4-dihydroisoquinoline;hydrochloride
2724664
98%
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : NW7625000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only