Please login to your online account to display your discounted pricing

6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  BTB04055DA

 View more versions of this product

Catalog No. BTB04055DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.26
Quantity 1g
Chemical Name or Material 6, 7-Dimethoxy-1-methyl-3, 4-dihydroisoquinoline
Assay Percent Range 97%
Molecular Formula C12H15NO2
CAS 4721-98-6

Specifications

Chemical Name or Material 6, 7-Dimethoxy-1-methyl-3, 4-dihydroisoquinoline
Assay Percent Range 97%
Molecular Formula C12H15NO2
Formula Weight 205.26