Please login to your online account to display your discounted pricing

6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 6, 7-Dimethoxy-1-methyl-3, 4-dihydroisoquinoline
Assay Percent Range 97%
Molecular Formula C12H15NO2
Formula Weight 205.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB04055DA

  SDS
fisher scientific company
BTB04055DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.26
Quantity 1g
Assay Percent Range 97%
CAS 4721-98-6
Chemical Name or Material 6, 7-Dimethoxy-1-methyl-3, 4-dihydroisoquinoline
Molecular Formula C12H15NO2